missing translation for 'onlineSavingsMsg'
Learn More

(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%

Product Code. 11955201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11955201 5 g Glass bottle 5g
11965201 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11955201 Supplier Thermo Scientific Acros Supplier No. 439540050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69460-11-3 | C10H19N | 153.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69460-11-3
Molecular Formula C10H19N
Molecular Weight (g/mol) 153.27
InChI Key VPTSZLVPZCTAHZ-BZNPZCIMSA-N
Synonym 1r,2r,3r,5s---isopinocampheylamine, 1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine, bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s, --isopinocampheylamine, --isopinocampheyl amine, 1r,2r,3r,5s-3-pinanamine, 1r,2r,5s-pinane-3alpha-amine, --isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine, 1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine
PubChem CID 11744832
IUPAC Name (1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
SMILES CC1C2CC(C2(C)C)CC1N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 90°C (18.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -42
Formula Weight 153.27
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Show More Show Less
compliance-icons
Product Identifier
  • (1R,2R,3R,5S)-(-)-Isopinocampheylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.