CAS RN 69460-11-3
IUPAC Name:
2,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
Synonyms:
2,6,6-trimethylbicyclo[3.1.1]heptan-3-amine
Molecular Weight (g/mol):
153.27
Molecular Formula:
C10H19N
InChi Key:
VPTSZLVPZCTAHZ-UHFFFAOYNA-N
SMILES:
CC1C2CC(CC1N)C2(C)C
1
–
1
of
1
results
1
(1R,2R,3R,5S)-(-)-Isopinocampheylamine, 95%
CAS: 69460-11-3 Molecular Formula: C10H19N Molecular Weight (g/mol): 153.27 InChI Key: VPTSZLVPZCTAHZ-BZNPZCIMSA-N Synonym: 1r,2r,3r,5s---isopinocampheylamine,1r,2r,3r,5s-2,6,6-trimethylbicyclo 3.1.1 heptan-3-amine,bicyclo 3.1.1 heptan-3-amine, 2,6,6-trimethyl-, 1r,2r,3r,5s,--isopinocampheylamine,--isopinocampheyl amine,1r,2r,3r,5s-3-pinanamine,1r,2r,5s-pinane-3alpha-amine,--isopinocampheylamine; 1r,2r,3r,5s-3-pinanamine,1r,2r,3r,5s-2,6,6-trimeth-ylbicyclo 3.1.1 heptane-3-amine PubChem CID: 11744832 IUPAC Name: (1S,3R,4R,5R)-4,6,6-trimethylbicyclo[3.1.1]heptan-3-amine SMILES: CC1C2CC(C2(C)C)CC1N