missing translation for 'onlineSavingsMsg'
Learn More

(1R)-10-Camphorsulfonamide, 97%, Thermo Scientific™

Product Code. 11478510
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72597-34-3
Molecular Formula C10H17NO3S
Molecular Weight (g/mol) 231.31
MDL Number MFCD00151500
InChI Key SBLUNABTQYDFJM-MHPPCMCBSA-N
Synonym 1r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptan-1-yl methanesulfonamide, 1r---10-camphorsulfonamide, 1s-+-10-camphorsulfonamide, 1r-7,7-dimethyl-2-oxobicyclo 2.2.1 heptane-1-methanesulfonamide
PubChem CID 12840037
IUPAC Name [(4R)-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonamide
SMILES CC1(C2CCC1(C(=O)C2)CS(=O)(=O)N)C

Specifications

Melting Point 134°C (decomposition)
Quantity 1 g
Formula Weight 231.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (1R)-10-Camphorsulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'