accessibility menu, dialog, popup

UserName

CAS RN 72597-34-3

H3CH3C(S)(R)SH2NOOO

CAS RN 72597-34-3

IUPAC Name: [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonamide
Synonyms: [(1R,4S)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonamide
Molecular Weight (g/mol): 231.31
Molecular Formula: C10H17NO3S
InChi Key: SBLUNABTQYDFJM-XVKPBYJWSA-N
SMILES: CC1(C)[C@H]2CC[C@]1(CS(N)(=O)=O)C(=O)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results