missing translation for 'onlineSavingsMsg'
Learn More

1H-Naphtho[2,3-d][1,2,3]triazole, TRC

Product Code. 30474019
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30474019 1 g
30470287 100 mg
30472295 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474019 Supplier Toronto Research Chemicals Supplier No. TRCN6236531G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 269-12-5
Molecular Formula C10H7N3
Molecular Weight (g/mol) 169.18
Synonym 1,2,3-Triaza-5,6-benzindene, 2H-Benzo[f]benzotriazole
IUPAC Name 2H-benzo[f]benzotriazole
SMILES c1ccc2cc3n[nH]nc3cc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H7N3/c1-2-4-8-6-10-9(11-13-12-10)5-7(8)3-1/h1-6H,(H,11,12,13)
Formula Weight 169.064
Percent Purity >95%
Chemical Name or Material 1H-Naphtho[2,3-d][1,2,3]triazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.