CAS RN 269-12-5
CAS RN 269-12-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1H-Naphtho[2,3-d][1,2,3]triazole, TRC
CAS: 269-12-5 Molecular Formula: C10H7N3 Molecular Weight (g/mol): 169.18 Synonym: 1,2,3-Triaza-5,6-benzindene,2H-Benzo[f]benzotriazole IUPAC Name: 2H-benzo[f]benzotriazole SMILES: c1ccc2cc3n[nH]nc3cc2c1