Learn More
Agilent BioTek 192 Tube Wash Manifold for 384-well Microplates
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
406 FX Washer Dispenser
Unit Size:
Each
Description
- Washes 384-well plates two times faster than standard 96 tube manifold included with 406 FX Washer Dispenser
Specifications
Specifications
| Type | Manifold |
| For Use With (Equipment) | 406 FX Washer Dispenser |