missing translation for 'onlineSavingsMsg'
Learn More

19-Hydroxy Cholesterol, TRC

Product Code. 30394856
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394856 10 mg
30401276 1 mg
30396217 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394856 Supplier Toronto Research Chemicals Supplier No. TRCH91800510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 561-63-7
Molecular Formula C27H46O2
Molecular Weight (g/mol) 402.65
Synonym Cholest-5-ene-3,19-diol, (3β)-, Cholest-5-ene-3β,19-diol (8CI), (3β)-Cholest-5-ene-3,19-diol, 19-Hydroxycholesterol, 3β,19-Dihydroxycholest-5-ene, 19-Hydroxy cholesterol
IUPAC Name (3S,8S,9S,10S,13R,14S,17R)-10-(hydroxymethyl)-13-methyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(CO)[C@H]3CC[C@]12C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H46O2/c1-18(2)6-5-7-19(3)23-10-11-24-22-9-8-20-16-21(29)12-15-27(20,17-28)25(22)13-14-26(23,24)4/h8,18-19,21-25,28-29H,5-7,9-17H2,1-4H3/t19-,21+,22+,23-,24+,25+,26-,27-/m1/s1
Formula Weight 402.3498
Chemical Name or Material 19-Hydroxy Cholesterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.