CAS RN 561-63-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
19-Hydroxy Cholesterol, TRC
CAS: 561-63-7 Molecular Formula: C27H46O2 Molecular Weight (g/mol): 402.65 Synonym: Cholest-5-ene-3,19-diol, (3β)-,Cholest-5-ene-3β,19-diol (8CI),(3β)-Cholest-5-ene-3,19-diol,19-Hydroxycholesterol,3β,19-Dihydroxycholest-5-ene,19-Hydroxy cholesterol IUPAC Name: (3S,8S,9S,10S,13R,14S,17R)-10-(hydroxymethyl)-13-methyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(CO)[C@H]3CC[C@]12C