missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ 18/10 Stainless Steel Weighing Scoop

Product Code. 11895071
Click to view available options
Length (Metric):
220 mm
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11895071

Brand: Bochem™ 360110

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Quantity 10 Pk.
Product Type Weighing Scoop
Diameter (Metric) Outer 5mm
Handle Material Stainless Steel (18/10)
Material Stainless Steel (18/10)
Length (Metric) 220 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.