Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrer
Brand:  Bochem™ 9012
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15256427
  • £33.30 / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Specifications
Propeller stirrer, 4 fix blades
8 mm
500 mm
70 mm
Mixing of media with low and middle viscosity