Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrer
Brand:  Bochem™ 9058
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10060781
  • £28.45 / Each
Estimated Shipment: 30-05-2024
to see stock.
Add to basket
Specifications
Propeller stirrer, 4 vertical blades
10 mm
500 mm
100 mm
Mixing of media with low and middle viscosity