missing translation for 'onlineSavingsMsg'
Learn More

17-Beta-Estradiol Antibody, Novus Biologicals™

Sheep Polyclonal Antibody

£160.00 - £381.00

Specifications

Antigen 17-Beta-Estradiol
Concentration 5.0 mg/mL
Dilution ELISA 1:5000-1:40000
Applications ELISA
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18071314
missing translation for 'viewDocuments'
Novus Biologicals
NB100-62282-0.05ML
0.05 mL
£160.00
0.05mL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
18064575
missing translation for 'viewDocuments'
Novus Biologicals
NB100-62282
0.25 mL
£381.00
0.25mL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

17-Beta-Estradiol Polyclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications

Specifications

17-Beta-Estradiol
ELISA 1:5000-1:40000
Polyclonal
Purified
RUO
PBS with 0.09% Sodium Azide
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
5.0 mg/mL
ELISA
Unconjugated
Sheep
All species
IgG
Protein G purified
NB100-62282 recognizes 17 beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Oestradiol 100% Oestrone 14% Oestriol 5% Aldosterone, Cortisol, Corticosterone, DHEA, Deoxycorticosterone, Pregnenolone, Progesterone, 17OH-P and Testosterone all <0.01%
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.