missing translation for 'onlineSavingsMsg'
Learn More
Learn More
17-Beta-Estradiol Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62282
This item is not returnable.
View return policy
Description
17-Beta-Estradiol Polyclonal specifically detects 17-Beta-Estradiol in All Species samples. It is validated for ELISA.
Specifications
| 17-Beta-Estradiol | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.09% Sodium Azide | |
| Sheep | |
| Protein G purified | |
| RUO | |
| NB100-62282 recognizes 17 beta estradiol, a mammalian estrogenic hormone, produced in the ovaries, placenta and testis that has a role in normal sexual development and the regulation of the menstrual cycle. The following cross reactivites have been observed: 17-beta Oestradiol 100% Oestrone 14% Oestriol 5% Aldosterone, Cortisol, Corticosterone, DHEA, Deoxycorticosterone, Pregnenolone, Progesterone, 17OH-P and Testosterone all <0.01% | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| 5.0 mg/mL | |
| ELISA 1:5000-1:40000 | |
| 17 beta Oestradiol, 17-Beta-Estradiol, 17-beta-HSD | |
| 17-beta estradiol conjugated to bovine serum albumin at position 6. | |
| 0.25 mL | |
| Primary | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction