missing translation for 'onlineSavingsMsg'
Learn More

17-(Allylamino)-17-demethoxygeldanamycin, 99%

Product Code. 15438539
Change view
Click to view available options
Quantity:
25 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438539 100 mg 100mg
15428539 25 mg 25mg
15408539 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438539 Supplier Thermo Scientific Alfa Aesar Supplier No. J66960.MC
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Possesses anti-tumor activity | CAS: 75747-14-7 | C31H43N3O8 | 585.698 g/mol

17-(Allylamino)-17-demethoxygeldanamycin (17-AAG) inhibits heat shock protein 90 (Hsp90), the expression of heat shock factor-1, and the activity of oncogenic proteins such as N-ras, Ki-ras, c-Akt, and p185erB2. In addition to its anti-tumor effects, 17-AAG also induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in DMSO at 150 mg/mL; soluble in ethanol at 5 mg/mL. Very poorly soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75747-14-7
Molecular Formula C31H43N3O8
Molecular Weight (g/mol) 585.698
MDL Number MFCD04973892
InChI Key AYUNIORJHRXIBJ-MKJFUHJFSA-N
Synonym 17-aag
PubChem CID 133688938
IUPAC Name [(3S,5R,6S,7R,8Z,11R,14E)-6-hydroxy-5,11-dimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-21-(prop-2-enylamino)-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
SMILES CC1CC(C(C(C=C(C(C(C=CC=C(C(=O)NC2=CC(=O)C(=C(C1)C2=O)NCC=C)C)OC)OC(=O)N)C)C)O)OC

Specifications

Melting Point 207°C to 209°C
Flash Point >110°C (230°F)
Quantity 100 mg
Sensitivity Light sensitive
Solubility Information Soluble in DMSO at 150 mg/mL; soluble in ethanol at 5 mg/ml; Very poorly soluble in water
Formula Weight 585.69
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 17-(Allylamino)-17-demethoxygeldanamycin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.