missing translation for 'onlineSavingsMsg'
Learn More

Welch™ CRVpro™ Direct Drive Rotary Vane Vacuum Pump

The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.

Brand:  Welch™ 3042-00

Additional Details : Weight : 21.00000kg

 View more versions of this product

Product Code. 16228250

  • £2045.80 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Welch Gardner Denver CRVpro Two-Stage Rotary Vane Vacuum Pump
  • The robust vacuum pump CRVpro belongs to the new high-performance two-stage rotary vane pumps generation of Welch.
  • The pump impresses with its long service life, absolute reliability and its low maintenance requirements.
  • This has been enabled thanks to 3 major innovations:
    • Its low running temperature, which slows down potential corrosion processes and reduces the oil consumption.
    • Its large oil reservoir ensures a low chemical concentration, which contributes to the prolongation of the oil exchange intervals.
    • Its special inside coating increases the chemical resistance of the pump.
  • Scope of delivery: Pump with motor, 3 line cords (EU/UK/US), 2 liters of oil, centering ring with dirt trap, centering ring, 2 clamping rings.
Specifications

Specifications

0.5 hp
1450 rpm
18.2 x 6.1 x 9.1 in.
46.3 lb.
10°C
Rotary Vane Vacuum Pump
1150 mL
230 V
Pump with Motor, 3 Line Cords (EU/UK/US), 2 L of Oil, Centering Ring with Dirt Trap, Centering Ring, 2 Clamping Rings
cUL and CE
CRVpro 4
40°C
50/60 Hz
2.8 CFM, 78 L/min.
2 x 10-3 mbar (1.5 x 10-3 torr)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers