Learn More
Agilent BioTek 1536-well Microplate Wash Aspiration Manifold Kit
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
406 FX Washer Dispenser
Unit Size:
Each
Description
- Includes aspiration manifold, alignment jig and instructions
- Aspirate-only manifold is used with either a dual syringe pump dispenser module and 32-tube dispense manifold(s), or with a peristaltic pump dispenser module and a 1536-well microplate cassette
- Dispense manifolds and cassettes are ordered separately
- For syringe pump dispensing into 1536-well plates, at least one 32-tube dispense manifold is required
- For faster dispensing, two 32-tube dispense manifolds can be used simultaneously
Specifications
Specifications
| Type | Wash Module |
| For Use With (Equipment) | 406 FX Washer Dispenser |