missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 15-PGDH Recombinant Rabbit Monoclonal Antibody (025)

Rabbit Recombinant Monoclonal Antibody
£91.00 - £357.00
Specifications
| Antigen | 15-PGDH |
|---|---|
| Clone | 025 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30535745
![]() |
Invitrogen™
MA560316 |
100 μL |
£357.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30535762
|
Invitrogen™
MA560317 |
20 μL |
£91.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
15-PGDH Recombinant Monoclonal Antibody for ELISA
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.Specifications
| 15-PGDH | |
| 1 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| P15428 | |
| 3248 | |
| Recombinant Human HPGD/15-PGDH protein (Met1-Gln266). | |
| Antibody |
| 025 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| HPGD | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; AV026552; HPGD; hydroxyprostaglandin dehydrogenase 15 (NAD); hydroxyprostaglandin dehydrogenase 15-(NAD); NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; NAD-dependent 15-hydroxyprostaglandin dehydrogenase; PGDH; PGDH1; PHOAR1; Prostaglandin dehydrogenase 1; SDR36C1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1 | |
| HPGD | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title