Learn More
Invitrogen™ 15-PGDH Recombinant Rabbit Monoclonal Antibody (025)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560316
 View more versions of this product
Product Code. 30535745
  • £357.00 / 100µL
Description
15-PGDH Recombinant Monoclonal Antibody for ELISA
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15-PGDH
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P15428
HPGD
Recombinant Human HPGD/15-PGDH protein (Met1-Gln266).
100 μL
Primary
Human
Antibody
IgG
ELISA
025
Unconjugated
HPGD
15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; AV026552; HPGD; hydroxyprostaglandin dehydrogenase 15 (NAD); hydroxyprostaglandin dehydrogenase 15-(NAD); NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; NAD-dependent 15-hydroxyprostaglandin dehydrogenase; PGDH; PGDH1; PHOAR1; Prostaglandin dehydrogenase 1; SDR36C1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1
Rabbit
Protein A
RUO
3248
-20°C, Avoid Freeze/Thaw Cycles