missing translation for 'onlineSavingsMsg'
Learn More
Learn More
15-Lipoxygenase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00321-50ul
This item is not returnable.
View return policy
Description
15-Lipoxygenase 2 Polyclonal antibody specifically detects 15-Lipoxygenase 2 in Human samples. It is validated for ELISA
Specifications
| 15-Lipoxygenase 2 | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| 15-lipoxygenase 2, 15-LOX-B, arachidonate 15-lipoxygenase 2, arachidonate 15-lipoxygenase B, Arachidonate 15-lipoxygenase type II, arachidonate 15-lipoxygenase, second type, arachidonate 15-lipoxygenase, type B, arachidonate omega(6) lipoxygenase, EC 1.13.11, EC 1.13.11.33,15-LOX-215S-lipoxygenase | |
| Produced in rabbits immunized with purified, recombinant Human 15-Lipoxygenase 2 (Accession#: AAH35217.1; Met1-Ile676). | |
| 50 μg | |
| Lipid and Metabolism | |
| 247 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction