missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E10A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66061-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
66061-1-Ig recognizes 14-3-3 tau (YWHAQ), eta (YWHAH), and zeta (YWHAZ), but not epsilon (YWHAE).
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5′ UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.Specifications
14-3-3 | |
Monoclonal | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P27348, P68254, P68255 | |
Ywhaq | |
14-3-3 Fusion Protein Ag6055 | |
20 μL | |
Primary | |
Porcine, Mouse, Rat, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Flow Cytometry, Immunofluorescence | |
2E10A3 | |
Unconjugated | |
YWHAQ | |
14 3 3, 14 3 3 protein T cell, 14 3 3 protein tau, 14 3 3 protein theta, 14-3-3 tau, HS1, Protein HS1, YWHAQ | |
Mouse | |
Protein G | |
RUO | |
100048964, 10971, 22630, 25577 | |
-20°C | |
Liquid |