missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2E10A3, Proteintech
Mouse Monoclonal Antibody
£130.00 - £339.00
Specifications
Antigen | 14-3-3 |
---|---|
Clone | 2E10A3 |
Concentration | 0.71 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16877595
|
Proteintech
66061-1-IG-20UL |
20 μL |
£130.00
20µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16867595
|
Proteintech
66061-1-IG-150UL |
150 μL |
£339.00
150µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
66061-1-Ig recognizes 14-3-3 tau (YWHAQ), eta (YWHAH), and zeta (YWHAZ), but not epsilon (YWHAE).
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5′ UTR a 6 bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease.Specifications
14-3-3 | |
0.71 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
14 3 3, 14 3 3 protein T cell, 14 3 3 protein tau, 14 3 3 protein theta, 14-3-3 tau, HS1, Protein HS1, YWHAQ | |
Ywhaq | |
IgG1 | |
Protein G | |
Antibody |
2E10A3 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Porcine, Rat | |
P27348, P68254, P68255 | |
100048964, 10971, 22630, 25577 | |
14-3-3 Fusion Protein Ag6055 | |
Primary | |
-20°C | |
YWHAQ |