missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TBX Medium

Product Code. 13235439
Change view
Click to view available options
Fill Weight:
99-101 g
Format:
125 mL sirop bottle
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
13235439 99-101 g 125 mL sirop bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13235439 Supplier Thermo Scientific™ Supplier No. BO0194M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect E. coli in foods using this medium.

Tryptone Bile Glucuronide (TBX) Medium is based on Tryptone Bile Agar, which was originally formulated to improve on earlier methods used to detect E. coli in foods in terms of speed, reliability, better recovery from frozen samples and the detection of poor lactose fermenters

  • Tryptone Bile Glucuronide (TBX) Medium is based on Tryptone Bile Agar, which was originally formulated to improve on earlier methods used to detect E. coli in foods in terms of speed, reliability, better recovery from frozen samples and the detection of poor lactose fermenters
TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2 to 10°C
Format 125 mL sirop bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 99-101 g
pH 10 sirop bottles with screw cap closures in a box.
Quantity 10 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.