missing translation for 'onlineSavingsMsg'
Learn More

13-Ethyl-3-ethynyl-18,19-dinor-17Alpha-pregna-3,5-dien-20-yn-17-ol(Levo Norgestrel Impurity), TRC

Product Code. 30380398
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380398 25 mg -
30369193 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380398 Supplier Toronto Research Chemicals Supplier No. TRCE91788025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1337972-89-0
Molecular Formula C23 H28 O
Molecular Weight (g/mol) 320.47
Synonym 18,19-Dinorpregna-3,5-dien-20-yn-17-ol, 13-ethyl-3-ethynyl-, (17α)-, Levonorgestrel Imp. C (EP)
IUPAC Name (8R,9S,10R,13S,14S,17R)-13-ethyl-3,17-diethynyl-2,7,8,9,10,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ol
SMILES CC[C@]12CC[C@H]3[C@@H](CC=C4C=C(CC[C@H]34)C#C)[C@@H]1CC[C@@]2(O)C#C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C23H28O/c1-4-16-7-9-18-17(15-16)8-10-20-19(18)11-13-22(5-2)21(20)12-14-23(22,24)6-3/h1,3,8,15,18-21,24H,5,7,9-14H2,2H3/t18-,19+,20+,21-,22-,23-/m0/s1
Formula Weight 320.214
Chemical Name or Material 13-Ethyl-3-ethynyl-18,19-dinor-17a-pregna-3,5-dien-20-yn-17-
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.