CAS RN 1337972-89-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
13-Ethyl-3-ethynyl-18,19-dinor-17Alpha-pregna-3,5-dien-20-yn-17-ol(Levo Norgestrel Impurity), TRC
CAS: 1337972-89-0 Molecular Formula: C23 H28 O Molecular Weight (g/mol): 320.47 Synonym: 18,19-Dinorpregna-3,5-dien-20-yn-17-ol, 13-ethyl-3-ethynyl-, (17α)-,Levonorgestrel Imp. C (EP) IUPAC Name: (8R,9S,10R,13S,14S,17R)-13-ethyl-3,17-diethynyl-2,7,8,9,10,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ol SMILES: CC[C@]12CC[C@H]3[C@@H](CC=C4C=C(CC[C@H]34)C#C)[C@@H]1CC[C@@]2(O)C#C