missing translation for 'onlineSavingsMsg'
Learn More

13-cis-Fenretinide, TRC

Product Code. 30394129
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394129 1 g
30389718 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394129 Supplier Toronto Research Chemicals Supplier No. TRCF2500051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75686-07-6
Molecular Formula C26 H33 N O2
Molecular Weight (g/mol) 391.55
Synonym 13-cis-N-(4-Hydroxyphenyl)retinamide, WH 13
IUPAC Name (2Z,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide
SMILES C\C(=C\C(=O)Nc1ccc(O)cc1)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C26H33NO2/c1-19(11-16-24-21(3)10-7-17-26(24,4)5)8-6-9-20(2)18-25(29)27-22-12-14-23(28)15-13-22/h6,8-9,11-16,18,28H,7,10,17H2,1-5H3,(H,27,29)/b9-6+,16-11+,19-8+,20-18-
Formula Weight 391.2511
Chemical Name or Material 13-cis-Fenretinide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.