CAS RN 75686-07-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
13-cis-Fenretinide, TRC
CAS: 75686-07-6 Molecular Formula: C26 H33 N O2 Molecular Weight (g/mol): 391.55 Synonym: 13-cis-N-(4-Hydroxyphenyl)retinamide,WH 13 IUPAC Name: (2Z,4E,6E,8E)-N-(4-hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenamide SMILES: C\C(=C\C(=O)Nc1ccc(O)cc1)\C=C\C=C(/C)\C=C\C2=C(C)CCCC2(C)C