missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 11mm PE Snap Ring Seal, Red, Center hole, assembled septum

Product Code. 11797587 Shop All Fisherbrand Products
Click to view available options
Color:
Blue, White, Red
Red, White
Red-Orange, Transparent, Red
Material (Liner):
Natural Rubber/TEF
Silicone/PTFE
Thickness (Metric):
1 mm
1.3 mm
Unit Size:
Pack of 1000

Product Code. 11797587

Only null left
Add to basket
Add to basket

Polyethylene caps are chemically inert and suitable for most chromatography applications.

  • Redesign of locking tabs provide easier application and removal of caps
  • Enlarged open area allows for needle penetration across the entire vial opening
  • Textured outer surface for easier gripping and improved detection by autosamplers with optical vial sensors
  • Fully compatible with all Snap-It vials
  • Eliminate the need for crimping or de-capping tools
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
TRUSTED_SUSTAINABILITY

Specifications

Description Hard
Material (Liner) Silicone/PTFE
Color Blue, White, Red
Diameter (Metric) Outer 11 mm
Material Polyethylene
Pre-slit Yes
For Use With (Application) HPLC
Quantity 100
Thickness (Metric) 1 mm
Type Snap Ring Cap
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.