missing translation for 'onlineSavingsMsg'
Learn More

11-Keto Loteprednol Etabonate, TRC

Product Code. 30390039
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390039 500 mg
30396124 100 mg
30393797 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390039 Supplier Toronto Research Chemicals Supplier No. TRCK195405500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207670-54-0
Molecular Formula C24H29ClO7
Molecular Weight (g/mol) 464.94
Synonym (8S,9S,10R,13S,14S,17R)-Chloromethyl 17-((Ethoxycarbonyl)oxy)-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carboxylate
IUPAC Name chloromethyl (8S,9S,10R,13S,14S,17R)-17-ethoxycarbonyloxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylate
SMILES O=C1C=C[C@@]2(C)C(CC[C@]([C@@]3([H])[C@]([C@](C(OCCl)=O)(OC(OCC)=O)CC3)(C)C4)([H])[C@]2([H])C4=O)=C1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C24H29ClO7/c1-4-30-21(29)32-24(20(28)31-13-25)10-8-17-16-6-5-14-11-15(26)7-9-22(14,2)19(16)18(27)12-23(17,24)3/h7,9,11,16-17,19H,4-6,8,10,12-13H2,1-3H3/t16-,17-,19+,22-,23-,24-/m0/s1
Formula Weight 464.1602
Chemical Name or Material 11-Keto Loteprednol Etabonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.