CAS RN 207670-54-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
11-Keto Loteprednol Etabonate, TRC
CAS: 207670-54-0 Molecular Formula: C24H29ClO7 Molecular Weight (g/mol): 464.94 Synonym: (8S,9S,10R,13S,14S,17R)-Chloromethyl 17-((Ethoxycarbonyl)oxy)-10,13-dimethyl-3,11-dioxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carboxylate IUPAC Name: chloromethyl (8S,9S,10R,13S,14S,17R)-17-ethoxycarbonyloxy-10,13-dimethyl-3,11-dioxo-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylate SMILES: O=C1C=C[C@@]2(C)C(CC[C@]([C@@]3([H])[C@]([C@](C(OCCl)=O)(OC(OCC)=O)CC3)(C)C4)([H])[C@]2([H])C4=O)=C1