missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 10X TdT Stop Buffer

Product Code. 18748276
Click to view available options
Quantity:
20 mL
Unit Size:
20mL
This item is not returnable. View return policy

Product Code. 18748276

Brand: R&D Systems™ 48176003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ 10X TdT Stop Buffer is provided with the TACS™ TdT kits to stop the activity of the TdT enzyme in TUNEL labeling assays.

  • Long Name: Terminal Deoxynucleotidyl Transferase
  • Do not use past expiration date
  • There are no hazardous ingredients as defined in the United States 29 CFR 1910.1200 (OSHA) or the Canadian Controlled Products Regulations (WHMIS) and therefore not subject to the labelling requirements of the aforementioned standards.
  • Exposure may aggravate those with pre-existing eye, skin, or respiratory conditions
  • Store in a dry, cool, and well-ventilated place
  • Keep container closed when not in use
  • Keep/Store away from direct sunlight, extremely high or low temperatures, and incompatible materials
  • Uncontrolled product according to WHMIS classification criteria
TRUSTED_SUSTAINABILITY

Specifications

Quantity 20 mL
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.