missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences 10x Buffer CI
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
85mL
Description
- 10x Buffer CI
- 10x concentrated buffer suitable for cell isolation
- Contains 10x PBS with 10 mM EDTA and 5% BSA
Specifications
Specifications
| Formulation | 10 x PBS with 10 mM EDTA and 5% BSA |
| Form | Liquid |
| For Use With (Application) | Buffer for Cell Isolation and Staining |
| Quantity | 1 Ea. |
Safety and Handling
Product Identifier
- Buffer CI (10x) for cell isolation
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : sodium azide
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction