missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRP4 Recombinant Rabbit Monoclonal Antibody (23GB4950), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552742
This item is not returnable.
View return policy
Description
Secreted Frizzled-Related Proteins (sFRPs) are a family of glycosylated Wnt antagonists characterized by a conserved cysteine-rich domain that shares homology with the cysteine-rich, extracellular domain Frizzled proteins use for the binding of Wnt proteins and receptors. Lacking the transmembrane and intracellular domains of the Frizzled proteins, sFRPs function as soluble modulators of the Wnt signaling pathway through the direct binding of Wnt proteins to this cysteine-rich domain, and the resultant inhibition of Wnt receptor binding and signaling capabilities. sFRP-4 is widely distributed in a variety of embryonic and adult tissues where it can function as a circulating antiangiogenic factor, a potent proapoptotic factor, an inhibitor of insulin secretion, and a suppressor of both tumor growth and metastatic potential through disruption of the Wnt signaling pathway. Research has demonstrated the existence of a direct correlation between the downregulation and/or absence of circulating sFRP-4 and the progression of several cancer types, including ovarian, endometrial, prostate and lung. Upregulation of circulating sFRP-4 has been linked to the deterioration of glucose metabolism in the case of type 2 diabetes, as well as the suppression of the keratinocyte hyperproliferation and epidermal hyperlasia that are definitive of psoriasis.
Specifications
| SFRP4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SFRP4 | |
| Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20379, 6424, 89803 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4950 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6FHJ7, Q9JLS4, Q9Z1N6 | |
| SFRP4 | |
| A synthesized peptide derived from human SFRP4 (251-300AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction