missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRP4 Recombinant Rabbit Monoclonal Antibody (23GB4950), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | SFRP4 |
|---|---|
| Clone | 23GB4950 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089114
![]() |
Invitrogen™
MA552742 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088485
|
Invitrogen™
MA552743 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFRP4 Recombinant Monoclonal antibody specifically detects SFRP4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| SFRP4 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q6FHJ7, Q9JLS4, Q9Z1N6 | |
| 20379, 6424, 89803 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4950 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4 | |
| A synthesized peptide derived from human SFRP4 (251-300AA). | |
| Primary | |
| -20°C | |
| SFRP4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title