missing translation for 'onlineSavingsMsg'
Learn More

GPRC5D Antibody (3D4), Novus Biologicals™

Human Monoclonal Antibody

Brand:  Novus Biologicals NBP3-26547-100ul

 View more versions of this product

Product Code. 30043593

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPRC5D
Monoclonal
Unconjugated
50% Glycerol, 0.01M PBS, PH 7.4
Human
Affinity purified
RUO
Primary
Human
Purified
ELISA
3D4
ELISA
G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor
Recombinant Human GPRC5D protein [UniProt Q9NZD1]
100 μL
GPCR
55507
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.