missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGCR8 Recombinant Rabbit Monoclonal Antibody (23GB1615), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553980
This item is not returnable.
View return policy
Description
This gene encodes a subunit of the microprocessor complex which mediates the biogenesis of microRNAs from the primary microRNA transcript. The encoded protein is a double-stranded RNA binding protein that functions as the non-catalytic subunit of the microprocessor complex. This protein is required for binding the double-stranded RNA substrate and facilitates cleavage of the RNA by the ribonuclease III protein, Drosha. Alternate splicing results in multiple transcript variants.
Specifications
| DGCR8 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DGCR8 | |
| C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 287954, 54487, 94223 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB1615 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8WYQ5, Q9EQM6 | |
| DGCR8 | |
| A synthesized peptide derived from human DGCR8 (400-773AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction