missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DGCR8 Recombinant Rabbit Monoclonal Antibody (23GB1615), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | DGCR8 |
|---|---|
| Clone | 23GB1615 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088612
![]() |
Invitrogen™
MA553980 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087616
|
Invitrogen™
MA553981 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DGCR8 Recombinant Monoclonal antibody specifically detects DGCR8 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| DGCR8 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8WYQ5, Q9EQM6 | |
| 287954, 54487, 94223 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1615 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| C22orf12; D16H22S1742E; D16H22S788E; D16Wis2; DGCR8; DGCR8 microprocessor complex subunit; DGCR8, microprocessor complex subunit; DGCRK6; DiGeorge syndrome critical region 8; diGeorge syndrome critical region 8 homolog; DiGeorge syndrome critical region gene 8; Gy1; LP4941; Microprocessor complex subunit DGCR8; mir-1306; N41; pasha; sb:eu827; sb:eu832; si:ch211-106a19.4; Vo59c07; wu:fc23f08; wu:fc38f06 | |
| A synthesized peptide derived from human DGCR8 (400-773AA). | |
| Primary | |
| -20°C | |
| DGCR8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title