missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Antibody (SR1765), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21967-100ul
This item is not returnable.
View return policy
Description
CD83 Monoclonal antibody specifically detects CD83 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| CD83 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1765 | |
| Western Blot 1:500-1:2000 | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| A synthesized peptide derived from human CD83 (Uniprot #: Q01151) | |
| 100 μg | |
| Dendritic Cell Markers | |
| 9308 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction