missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD83 Antibody (SR1765), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | CD83 |
|---|---|
| Clone | SR1765 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18672825
|
Novus Biologicals
NBP3-21967-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615474
|
Novus Biologicals
NBP3-21967-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD83 Monoclonal antibody specifically detects CD83 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CD83 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| B-cell activation protein, BL11CD83 antigen, CD83 antigen (activated B lymphocytes, immunoglobulin superfamily), CD83 molecule, Cell surface protein HB15, cell-surface glycoprotein, HB15hCD83 | |
| A synthesized peptide derived from human CD83 (Uniprot #: Q01151) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1765 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Dendritic Cell Markers | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9308 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title