missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 5 Recombinant Rabbit Monoclonal Antibody (23GB3225), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552550
This item is not returnable.
View return policy
Description
Many isoforms of aquaporin have been identified in mammals, designated AQP0 through AQP10. Aquaporins are widely distributed and it is not uncommon for more than one type of AQP to be present in the same cell. Although most aquaporins are only permeable to water, AQP3, AQP7, AQP9 and one of the two AQP10 transcripts are also permeable to urea and glycerol. AQP2 is the only water channel that is activated by vasopressin to enhance water reabsorption in the kidney collecting duct. Aquaporins are involved in renal water absorption, generation of pulmonary secretions, lacrimation, and the secretion and reabsorption of cerebrospinal fluid and aqueous humor. In the lung, AQP5 is responsible for the majority of water transport across the apical membrane of type I alveolar epithelial cells.
Specifications
| Aquaporin 5 | |
| Recombinant Monoclonal | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9WTY4 | |
| AQP5 | |
| A synthetic peptide of human AQP5 (224-265AA). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3225 | |
| Unconjugated | |
| AQP5 | |
| AQP 5; Aqp5; AQP-5; aquaporin 5; Aquaporin5; aquaporin-5; PPKB | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11830 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction