missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aquaporin 5 Recombinant Rabbit Monoclonal Antibody (23GB3225), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Aquaporin 5 |
|---|---|
| Clone | 23GB3225 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089039
|
Invitrogen™
MA552550 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087442
|
Invitrogen™
MA552551 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aquaporin 5 Recombinant Monoclonal antibody specifically detects Aquaporin 5 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Aquaporin 5 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| Q9WTY4 | |
| 11830 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3225 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AQP 5; Aqp5; AQP-5; aquaporin 5; Aquaporin5; aquaporin-5; PPKB | |
| A synthetic peptide of human AQP5 (224-265AA). | |
| Primary | |
| -20°C | |
| AQP5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title