missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neutrophil Elastase/ELA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4517
This item is not returnable.
View return policy
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Mouse samples. It is validated for Western Blot.
Specifications
| Neutrophil Elastase/ELA2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q3UP87 | |
| ELANE | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neutrophil Elastase/ELA2 Met1-Arg260 Accession # Q3UP87 | |
| 100 μg | |
| Primary | |
| Detects mouse Neutrophil Elastase/ELA2 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 887105 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1991 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction