missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neutrophil Elastase/ELA2 Antibody, R&D Systems™

Rat Monoclonal Antibody
£184.00 - £426.00
Specifications
| Antigen | Neutrophil Elastase/ELA2 |
|---|---|
| Clone | 887105 |
| Concentration | LYOPH |
| Dilution | Western Blot 0.5 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761958
![]() |
R&D Systems
MAB4517-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700938
![]() |
R&D Systems
MAB4517 |
100 μg |
£426.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neutrophil Elastase/ELA2 Monoclonal specifically detects Neutrophil Elastase/ELA2 in Mouse samples. It is validated for Western Blot.Specifications
| Neutrophil Elastase/ELA2 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q3UP87 | |
| 1991 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Neutrophil Elastase/ELA2 Met1-Arg260 Accession # Q3UP87 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Neutrophil Elastase/ELA2 in direct ELISAs and Western blots. |
| 887105 | |
| Western Blot 0.5 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2, ELA2granulocyte-derived elastase, ELANE, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, Leukocyte Elastase, medullasin, NE, neutrophil elastase, PMN elastase, PMN-E, polymorphonuclear elastase, SCN1 | |
| ELANE | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title