missing translation for 'onlineSavingsMsg'
Learn More

10-Hydroxy-2-decenoic acid, TRC

Product Code. 30012369
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012369 250 mg 250mg
30006226 500 mg 500mg
17982025 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012369 Supplier Toronto Research Chemicals Supplier No. TRCH943213250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14113-05-4
Molecular Formula C10 H18 O3
Molecular Weight (g/mol) 186.25
Synonym (E)-10-Hydroxy-2-decenoic acid
IUPAC Name (E)-10-hydroxydec-2-enoic acid
SMILES OCCCCCCC\C=C\C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C10H18O3/c11-9-7-5-3-1-2-4-6-8-10(12)13/h6,8,11H,1-5,7,9H2,(H,12,13)/b8-6+
Formula Weight 186.1256
Percent Purity >95%
Chemical Name or Material (E)-10-Hydroxy-2-Decenoic Acid (10-Hda)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.