accessibility menu, dialog, popup

UserName

HOOHO(E)

CAS RN 14113-05-4

IUPAC Name: (2E)-10-hydroxydec-2-enoic acid
Synonyms: queen bee acid 10-hydroxydecenoic acid
Molecular Weight (g/mol): 186.25
Molecular Formula: C10H18O3
InChi Key: QHBZHVUGQROELI-SOFGYWHQSA-N
SMILES: OCCCCCCC\C=C\C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)