missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 10-DEBC hydrochloride
Description
Selective inhibitor of Akt/PKB. Inhibits IGF-1-stimulated phosphorylation and activation of Akt (complete inhibition at 2.5 μM), suppressing downstream activation of mTOR, p70 S6 kinase and S6 ribosomal protein. Shows no activity at PDK1, SGK1 or PI 3-kinase. Inhibits cell growth (IC50 ∽ 2-6 μM) and induces apoptosis in rhabdomyosarcoma cells.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 381.34 |
| Purity | >99% |
| Molecular Weight (g/mol) | 381.341 |
| Chemical Name or Material | 10-DEBC hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?