missing translation for 'onlineSavingsMsg'
Learn More

10-Camphorsulfonic Acid Ethyl Ester, TRC

Product Code. 30135194
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135194 10 g 10g
30009331 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135194 Supplier Toronto Research Chemicals Supplier No. TRCC17504010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108481-13-6
Molecular Formula C12H20O4S
Molecular Weight (g/mol) 260.35
Synonym 7,7-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Ethyl Ester (+/-)-10-Camphorsulfonic Acid Ethyl Ester
IUPAC Name ethyl (7,7-dimethyl-2-oxo-1-bicyclo[2.2.1]heptanyl)methanesulfonate
SMILES Fc1ccc(cc1)CNc2cc(N)c(N)cc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H20O4S/c1-4-16-17(14,15)8-12-6-5-9(7-10(12)13)11(12,2)3/h9H,4-8H2,1-3H3
Percent Purity >95%
Chemical Name or Material 10-Camphorsulfonic Acid Ethyl Ester
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.