CAS RN 108481-13-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
10-Camphorsulfonic Acid Ethyl Ester, TRC
CAS: 108481-13-6 Molecular Formula: C12H20O4S Molecular Weight (g/mol): 260.35 Synonym: 7,7-Dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic Acid Ethyl Ester (+/-)-10-Camphorsulfonic Acid Ethyl Ester IUPAC Name: ethyl (7,7-dimethyl-2-oxo-1-bicyclo[2.2.1]heptanyl)methanesulfonate SMILES: Fc1ccc(cc1)CNc2cc(N)c(N)cc2