Learn More
Fisherbrand™ 10-17mm Tube Rack
Incorporates two rotatable, hinged shelves to accommodate varying tube size. The multi-purpose Fisherbrand™ 10-17mm tube facilitates more comfortable pipetting and makes efficient use of limited space.
Brand:  Fisherbrand™
Additional Details : Weight : 0.35000kg
Product Code. 15728177
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • The 10-17 mm Tube Rack comes with FOUR interchangeable tube segments.
  • Ergonomic handles for ease of transporting the rack.
  • Molded grid on both sides of the shelves for quick sample identification and retrieval.
Specifications
Specifications
Autoclavable
Tubes (40)
7.7 cm
Purple, Natural
10.7 cm
17 mm