missing translation for 'onlineSavingsMsg'
Learn More

1-Triacontanol (>90% purity), TRC

Product Code. 30133075
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133075 5 g 5g
30006069 1 g 1g
17988214 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133075 Supplier Toronto Research Chemicals Supplier No. TRCT7670505G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-50-0
Molecular Formula C30 H62 O
Molecular Weight (g/mol) 438.81
Synonym 1-Triacontanol, 1-Triacontanol (6CI, 7CI, 8CI, 9CI, ACI), 1-Hydroxytriacontane, Melissyl alcohol, Miraculan, Myricyl alcohol, n-Triacontanol, NSC 402492, NSC 405588, Nutron, Prosopol, Tomatex, Triacontanol, Triacontyl alcohol, Ultria, Well-Bloom
IUPAC Name triacontan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C30H62O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31/h31H,2-30H2,1H3
Formula Weight 438.4801
Chemical Name or Material 1-Triacontanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.